Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1(H)-Naphthotriazol ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-N0051-M005 CDX-N0051-M025
102990-616EA 88.62 USD InStock
102990-616 102990-618
1(H)-Naphthotriazol ≥98%
1(H)-Naphthotriazol
Fluorescent probe. Used for the detection of nitrite, nitric oxide release and S-nitrosothiols (RSNOs). Activatable DNA cleaving agent.


Formula: C₁₀H₇N₃
MW: 169.2 g/mol
Storage Temperature: Freezer
CAS Number: 269-12-5
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Red to brown powder.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty