Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Opipramol ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-O0042-M010 CDX-O0042-M050
102990-694EA 88.62 USD InStock
102990-694 102990-696
Opipramol ≥98% (by HPLC)
Opipramol
Tricyclic antidepressant and anxiolytic compound. Acts as a high affinity sigma receptor agonist, primarily at the sigma1 subtype, but also at the sigma2 subtype with somewhat lower affinity. Shows a low to moderate antagonistic affinity for the D2, 5-HT2, H1, H2, and muscarinic acetylcholine receptors. H1 and H2 receptor antagonism account for its antihistamine effects, and muscarinic acetylcholine receptor antagonism is responsible for its anticholinergic properties.


Formula: C₂₃H₂₉N₃O
Storage Temperature: Freezer
CAS Number: 315-72-0
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White powder.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty