Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(6-Aminohexyl)rhodamine 6G-amide bis(trifluoroacetate) ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-R0021-M010 CDX-R0021-M050
102990-924EA 123.23 USD InStock
102990-924 102990-926
N-(6-Aminohexyl)rhodamine 6G-amide bis(trifluoroacetate) ≥95%
N-(6-Aminohexyl)rhodamine 6G-amide bis(trifluoroacetate)
N-(6-Aminohexyl)rhodamine 6G-amide bis(trifluoroacetate) is a rhodamine 6G-amide derivative useful as a bioreagent in fluorescence assays.


Formula: C₃₂H₄₀N₄O₂·C₄H₂F₆O₄
MW: 740.73 g/mol
Storage Temperature: Freezer
CAS Number: 1140505-40-3
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty