Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

A-674563 hydrochloride ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

SYN-1110-M001 SYN-1110-M005 SYN-1110-M010
102992-106EA 261.69 USD InStock
102992-106 102992-108 102992-110
A-674563 hydrochloride ≥95%
A-674563 hydrochloride
A-674563 is a potent, orally available Akt1 inhibitor with an IC(50) of 11nM. It also displays activity against PKA and CDK2 with IC(50) values of 16 and 46nM respectively.


Formula: C₂₂H₂₂N₄O·HCl
MW: 394.9 g/mol
Storage Temperature: Freezer
CAS Number: 552325-73-2
Order Now
Specification Test Results
Purity >95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty