Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SMI-4a ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste

SYN-1113-M001 SYN-1113-M005 SYN-1113-M010 SYN-1113-M050
102992-126EA 74.77 USD InStock
102992-126 102992-128 102992-130 102992-132
SMI-4a ≥95%
SMI-4a
SMI-4a is a novel, ATP-competitive inhibitor of Pim1 kinase (IC(50) of 17nM). It kills a wide range of both myeloid and lymphoid cell lines.


Formula: C₁₁H₆F₃NO₂S
Storage Temperature: Freezer
MDL Number: MFCD01152003
CAS Number: 327033-36-3
Order Now
Specification Test Results
Purity >95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty