Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GSK2606414 ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste

SYN-1201-M001 SYN-1201-M005 SYN-1201-M010 SYN-1201-M050
102992-754EA 134.31 USD InStock
102992-754 102992-756 102992-758 102992-760
GSK2606414 ≥95%
GSK2606414
GSK2606414 is an orally available, potent, and selective PERK inhibitor with IC(50) of 0.4nM, displaying at least 100-fold selectivity over the other EIF2AKs assayed.


Formula: C₂₄H₂₀F₃N₅O
Storage Temperature: Freezer
MDL Number: MFCD25976926
CAS Number: 1337531-36-8
Order Now
Specification Test Results
Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty