Colistin Sulphate (from Bacillus polymyxa)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-CN2-0065-M100 AG-CN2-0065-G001 AG-CN2-0065-G005
102987-208EA 41.54 USD InStock
102987-208 102987-210 102987-212
Colistin Sulphate (from Bacillus polymyxa)
Proteins and Peptides
Very potent polymyxin antibiotic
  • Antimicrobial
Has only one amino acid difference compared to polymyxin B. Targets the lipopolysaccharide (LPS) of the outer membrane of Gram-negative bacteria. Competitively displaces divalent cations (Ca2+ and Mg2+) from the negatively-charged phosphate groups LPS, causing disruption of the membrane. This increases the permeability of the cell envelope, leakage of cell contents, leading to apoptosis and cell death. Used against multidrug-resistant Gram-negative bacterial infections.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty