Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fulvic acid ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

AG-CN2-0135-M001 AG-CN2-0135-M005
102988-032EA 193.85 USD InStock
102988-032 102988-034
Fulvic acid ≥95%
Fulvic acid
Free radical scavenger and antioxidant. Potential compound to neutralize radioactive and toxic wastes and "heal" soils. Chelates and binds scores of minerals into a bioavailable form used by cells. Shown to incorporate into bone and cartilage of rats. Antibacterial. Antifungal. beta-Hexosaminidase release inhibitor. Anti-inflammatory. Tau fibrils aggregation inhibitor (Alzheimers Disease). Antiseptic. Interrupts the dimer formation of Abeta(17-42) peptides.


Formula: C₁₄H₁₂O₈
MW: 308.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09838488
CAS Number: 479-66-3
Order Now
Specification Test Results
Source/Host Isolated from Penicillium sp. strain FKP-0046.
Purity 95% (HPLC)
Appearance/Color Yellow solid.
Solubility Soluble in methanol, benzene or chloroform. Insoluble in water.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty