Wort Agar, HiMedia Laboratories
Supplier: PAESEL+LOREI GMBH&CO
GM129-2.5KG GM129-500G GM129-5KG
75804-106EA 1022.22 USD InStock
75804-106 75804-108 75804-110
Wort Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Wort agar is used for the cultivation and enumeration of yeasts.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty