Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Water, UltraPure for molecular biology, sterile filtered

Supplier: Bandelin Electronic GmbH &Co.KG

MB-010-1000 MB-010-0100
RLMB-010-1000EA 128.51 USD InStock
RLMB-010-1000 RLMB-010-0100
Water, UltraPure for molecular biology, sterile filtered
Water
Molecular Biology Grade Ultrapure water is suitable for molecular immunology, molecular biology and nucleic acid methodologies, when applicable. It is commonly used in laboratory to prepare buffers in biochemistry and molecular biology laboratories and also cell culture media for cell biology studies.

Molecular Biology Grade UltraPure Water consists of highly polished pharmaceutical grade deionized water treated with diethyl pyrocarbonate (DEPC). This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.


Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty