Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Forskolin (from Coleus forskohlii)

Supplier: E. Becker & Co. GmbH
Warning

0219066901 0219066905 0219066910 0219066925 0219066950
IC19066901EA 66.17 USD InStock
IC19066901 IC19066905 IC19066910 IC0219066925 IC0219066950
Forskolin (from Coleus forskohlii)
Forskolin
Forskolin is the major diterpene isolated from the Indian plant Coleus forskohlii. At low doses, forskolin is a positive inotropic agent in dogs, cats, spontaneously hypertensive and normal rats and in an isolated guinea pig heart (Langendorf) preparation. At higher doses, forskolin is a hypotensive and vasodilatory agent because of its function as a smooth muscle relaxant. No major side effects are observed at effective doses.

Forskolin's pharmacological activities are due to its activation of adenylate cyclase, resulting in increased intracellular cyclic AMP in most tissues and cells. The exact mechanism of forskolin's positive inotropic effect is unknown but may be related to a cyclic AMP-dependent increase in Na+ permeability that results in an indirect augmentation of calcium release. Forskolin, 7-o-Hemisuccinyl-7-deacetyl is a biologically active and useful derivative of forskolin. It has been attached to Sepharose as an affinity support for the purification of adenylate cyclase.

Forkolin is reported to demonstrate positive inotropic and heart rate-increasing activity at low doses in animals. Also functions as an antihypertensive and vasodilator at higher doses. Activates adenylcyclase in rat liver membranes.

Cell-permeable diterpenoid that possesses anti-hypertensive, positive inotropic, and adenylyl cyclase activating properties. Many of its biological effects are due to its activation of adenylyl cyclase and the resulting increase in intracellular cAMP concentration. Forskolin effects calcium currents and inhibits MAP kinase.

Soluble at 6 mg per ml in 95% ethanol and at 2,5 mg per 5 µl in anhydrous dimethylsulphoxide (DMSO).


Formula: C₂₂H₃₄O₇
Melting Pt: 228…230 °C (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00082317
CAS Number: 66575-29-9
EINECS: 266-410-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=995895c46cb56bbdebccfc689e3b2632c6e29733&vid=995895c46cb56bbdebccfc689e3b2632c6e29733&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty