PSB Broth Base, HiMedia Laboratories
Supplier: PAESEL+LOREI GMBH&CO
M941-500G M941-2.5KG M941-5KG
95022-502EA 214.96 USD InStock
95022-502 89226-886 89226-888
PSB Broth Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
PSB broth base is used for primary enrichment and enumeration of Yersinia enterocolitica from foods.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty