Brady® Labeled Lockout Hasp, Brady Worldwide
Supplier: Brady Worldwide
65962 65963
12222-136PK 192.42 USD InStock
12222-136 12222-138
Brady® Labeled Lockout Hasp, Brady Worldwide
Security Locks
Labeled Lockout Hasps combine the advantages of lockout tags and safety hasps in a single convenient product.
  • Permanent front- and back-side labels on these aluminum hasps include a write-on area that accepts pen, pencil, and permanent marker
  • Hasps allow multiple padlocks to secure a single lockout device
  • Equipping employees with the proper lockout tools and warning devices can save lives, reduce lost employee time, and cut insurance costs
  • Brady® is a world leader in lockout/tagout products and services offering industry expertise and a broad range of durable, easy-to-use lockout devices
These tough, aluminum hasps accommodate group lockouts as up to 6 workers can apply their individual locks.
Certificaciones: OSHA 1910.147.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty