Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chalcomycin (from Streptomyces bikiniensis) ≥96% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

BVT-0263-C500 BVT-0263-M001
102988-912EA 152.31 USD InStock
102988-912 102988-914
Chalcomycin (from Streptomyces bikiniensis) ≥96% (by HPLC)
Chalcomycin
Chalcomycin is a 16-membered macrolide antibiotic.

  • Macrolide antibiotic
  • Protein biosynthesis inhibitor
  • Anti-bacterial and anti-fungal

Broad spectrum antimicrobial. Antibacterial and antifungal. Protein biosynthesis inhibitor.

Beige solid

Soluble in DMSO or acetone


Storage Temperature: Freezer
CAS Number: 20283-48-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty