Human Recombinant Stem Cell Factor
Supplier: SAFE AND SURE NEDERLAND B.V.
201-804-0002 201-804-0010 201-804-0100
76001-912EA 206.01 USD InStock
76001-912 76001-914 76288-308
Human Recombinant Stem Cell Factor
Proteins and Peptides
Produced in E. coli. Non-glycosylated protein, containing 165 amino acids.
  • High purity
  • Carrier-free
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty