Human Recombinant Activin-A
Supplier: SAFE AND SURE NEDERLAND B.V.
201-809-0010 201-809-0100
76001-916EA 344.44 USD InStock
76001-916 76001-918
Human Recombinant Activin-A
Proteins and Peptides
Produced in E. coli. Non-glycosylated homodimer, containing two 117 amino acid chains.
  • Carrier-free
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty