Mouse Recombinant IL-12
201-815-0010 201-815-0100
76001-930EA 337 USD InStock
76001-930 76001-932
Mouse Recombinant IL-12
Proteins and Peptides
Produced in HEK293 cells. Glycosylated, disulfide-linked heterodimer. It contains one p35 subunit (35 kDa) and one p40 subunit (40 kDa), with a total of 506 amino acids.
  • Carrier-free
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty