Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PD-0173074 ≥99% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

CHM112-0005 CHM112-0010
76002-486EA 262.69 USD InStock
76002-486 76002-488
PD-0173074 ≥99% (by HPLC)
PD-0173074
Cell-permeable, potent inhibitor of FGF and VEGF receptors.

A cell-permeable compound that acts as a potent, ATP-competitive, and reversible inhibitor of FGF and VEGF receptors. PD 173074 inhibits PDGFR and c-Src only at much higher concentration. Shown to inhibit the autophosphorylation of endogenous FGFR1 and overexpressed VEGFR2.


Formula: C₂₈H₄₁N₇O₃
MDL Number: MFCD08705327
CAS Number: 219580-11-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty