Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PP242 hydrate ≥99% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

CHM132-0001 CHM132-0005
76003-030EA 91.56 USD InStock
76003-030 76003-032
PP242 hydrate ≥99% (by HPLC)
PP242 hydrate
Inhibitor of both mTORC1 and mTORC2

Inhibitor of the active site of mTOR kinase in both mTORC1 and mTORC2. pp242 has been shown to cause the death of certain human and murine leukemia cells more potently than rapamycin.

Induces cell death in human and murine leukemia cells.


Formula: C₁₆H₁₆N₆O·H₂O
CAS Number: 1092351-67-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty