Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

WH-4-023 ≥99% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

CHM145-0005 CHM145-0010
76003-078EA 130.8 USD InStock
76003-078 76003-080
WH-4-023 ≥99% (by HPLC)
WH-4-023
Potent and orally active Lck/Src inhibitor.


Formula: C₃₂H₃₆N₆O₄
MDL Number: MFCD28138072
CAS Number: 837422-57-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty