Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Motesanib 99 HPLC_ASSAY_METHOD

Supplier: SAFE AND SURE NEDERLAND B.V.

CHM147-0001 CHM147-0005
76003-086EA 69.76 USD InStock
76003-086 76003-088
Motesanib 99 HPLC_ASSAY_METHOD
Motesanib


Formula: C₂₂H₂₃N₅O
MW: 373.5 g/mol
MDL Number: MFCD10567689
CAS Number: 453562-69-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty