Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride ≥95% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

CHM157-0005 CHM157-0050
76003-112EA 70.9 USD InStock
76003-112 76003-114
N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride ≥95% (by HPLC)
N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride
EGFR tyrosine kinase auto-phosphorylation inhibitor

Erlotinib inhibits EGFR tyrosine kinase auto-phosphorylation. Studies in cell lines and enzyme assays have shown that erlotinib inhibits EGFR at concentrations significantly lower than those needed to inhibit c-src and v-abl. It has an IC50 of 2 nM against EGFR. It is >1000-fold more sensitive for EGFR than for c-Src or v-Abl.


Formula: C₂₂H₂₄ClN₃O₄
MW: 429.901 g/mol
MDL Number: MFCD07781272
CAS Number: 183319-69-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty