Human Recombinant NMNAT2 (from E. coli)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40A-0153T-C010 AG-40A-0153T-3010
102980-078EA 193.85 USD InStock
102980-078 102980-080
Human Recombinant NMNAT2 (from E. coli)
Proteins and Peptides
Nicotinamide mononucleotide adenylyltransferase 2 (NMNAT2) catalyzes the formation of NAD+ from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form nicotinic acid mononucleotide (NaMN) as a substrate but with lower efficiency. NMNAT2 also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD+. It is highly expressed in brain, in particular in cerebrum, cerebellum, occipital lobe, frontal lobe, temporal lobe and putamen. NMNAT2 is also found in the heart, skeletal muscle, pancreas and islets of Langerhans. NMNAT2 is essential for axon growth and survival. Its loss from injured axons may activate Wallerian degeneration (axon degeneration induced by nerve injury), whereas NMNAT overexpression rescues axons from degeneration.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty