Human Recombinant FGF-2 (from E. coli)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40B-0150-C010 AG-40B-0150-3010
102980-318EA 138.46 USD InStock
102980-318 102980-320
Human Recombinant FGF-2 (from E. coli)
Proteins and Peptides
FGF-2 (Basic fibroblast growth factor) belongs to the FGF superfamily that is made up of 22 different fibroblast growth factor genes. FGF-2 and FGF-1 (acidic FGF) are unique in that they do not follow the conventional signal sequence for secretion. FGF-2 is found in five isoforms via alternative initiation of translation. Signaling of FGF-2 occurs through the high-affinity tyrosine kinase receptors FGFR1-4. FGF-2 is a potent wide-spectrum mitogen whose overexpression is associated with immortalization and unregulated cell proliferation in many tumors. FGF-2 functions in angiogenesis, cell survival, tissue repair, embryonic development and inflammation. FGF-2 is widely used to maintain the self-renewal of human embryonic stem cells (ESCs) and to induce pluripotent stem cells. FGF-2 is an established neurogenic factor for proliferation and differentiation of multipotent neural stem cells both during development and in the adult mouse brain as well.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty