Human Recombinant DLK1 (from HEK293 Cells)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40B-0152-C010 AG-40B-0152-3010
102980-326EA 332.31 USD InStock
102980-326 102980-328
Human Recombinant DLK1 (from HEK293 Cells)
Proteins and Peptides
The transmembrane and secreted protein delta-like 1 homolog (DLK1; Protein delta homolog 1; Preadipocyte factor 1; Pref-1) belongs to the EGF-like family. It can be cleaved by a tumor necrosis factor-alpha converting enzyme (TACE) to release a soluble form. DLK1 plays important roles in regulating cell differentiation such as adipogenesis and osteogenesis. Membrane-bound DLK1 promotes hypertrophic myotube formation, whereas soluble DLK1 inhibits myotube formation. The soluble form of DLK1 inhibits preadipocyte differentiation, while membrane DLK1 represses preadipocyte proliferation. DLK1 is also associated with tumor invasion.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty