Human Recombinant FNDC5 (from HEK293 cells)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40B-0153-C010 AG-40B-0153-3010
102980-330EA 332.31 USD InStock
102980-330 102980-332
Human Recombinant FNDC5 (from HEK293 cells)
Proteins and Peptides
Irisin is a recently described exercise and PGC1-alpha-induced myokine secreted by skeletal muscle in mice and humans. Irisin has been shown to induce browning of white adipose tissue, but this role has been questioned and a new function in the control of mass and strength of cortical bone has been reported. The membrane fibronectin type III domain containing protein 5 (FNDC5) can be cleaved by an unknown protease to release the Irisin protein (aa 32-143 for human) and potentially also the FNDC5 (extracellular domain, aa 32-153 for human).
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty