Human Recombinant VEGF 165 (from E. coli)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40T-0043-C005 AG-40T-0043-C020
102980-394EA 249.23 USD InStock
102980-394 102980-396
Human Recombinant VEGF 165 (from E. coli)
Proteins and Peptides
Human vascular endothelial growth factor (VEGF 165) is produced as a homodimer. It binds to the FLT1/VEGFR1 and KDR/VEGFR2 receptors, heparan sulfate and heparin. VEGF 165 is a specific mitogen for vascular endothelial cells and a strong angiogenic factor in vivo. In addition to its action as a mitogen it is a potent vascular permeability factor (VPF) in vivo. It is also a chemoattractant molecule for monocytes and endothelial cells. It induces endothelial cell proliferation, promotes cell migration, inhibits apoptosis and induces permeabilization of blood vessels.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty