Human Recombinant CD278 (from CHO Cells)
Supplier: Van Der Voort Lucht en Milieu Amste
CHI-HF-220ICOS-C025 CHI-HF-220ICOS-C100
102981-528EA 249.23 USD InStock
102981-528 102981-530
Human Recombinant CD278 (from CHO Cells)
Proteins and Peptides
CD278, also called inducible costimulator (ICOS) or CRP1 (CD28-related protein-1), is a member of the growing CD28 family of immune costimulatory receptors. Other family members are CD28, CTLA-4 and PD-1. ICOS is expressed on most CD45RO+ cells. ICOS expression is upregulated within approximately 24-48 hours of activation on Th primed cells. B7-H2, a member of the B7 family of costimulatory ligands, has been identified as the ICOS ligand. The B7-H2/ICOS interaction appears to play roles in T cell dependent B cell activation and Th differentiation.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty