Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium dodecyl sulphate (SDS) ≥99%, white powder

Supplier: E. Becker & Co. GmbH
Danger

0219483125 0219483180 0219483183 0219483190 0219483191
IC0219483125EA 59.46 USD InStock
IC0219483125 IC0219483180 IC0219483183 IC0219483190 IC0219483191
Sodium dodecyl sulphate (SDS) ≥99%, white powder
Sodium dodecyl sulphate (SDS)
SDS is an detergent typically used to solubilize and denature proteins for electrophoresis. SDS has also been used in large-scale phenol extraction of RNA to promote the dissociation of protein from nucleic acids when extracting from biological material. SDS is used in Protein Electrophoresis & Blotting.

SDS is an anionic detergent and wetting agent that is effective in both acid and alkaline solutions. Most proteins bind SDS in a ratio of 1.4 grams SDS to 1 gram protein. The charges intrinsic to the protein become insignificant compared to the overall negative charge provided by the bound SDS. The charge to mass ratio is essentially the same for each protein and will migrate in the gel based only on protein size. The salt is of an organosulfate consisting of a 12-carbon tail attached to a sulfate group, giving the material the amphiphilic properties required of a detergent.


Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 180…210 °C
Density: 1.1 (20 °C)
Flash Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 2296
ADR: 4.1 II
Merck Index: 14,08636
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8277ce6a5293059a472fc8e879ddad557808e93d&vid=8277ce6a5293059a472fc8e879ddad557808e93d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity Test Passes
Purity ~99%
FTIR Conforms to Standard
Melting Point 182 - 210 °C
OD260 nm (1% aq soln) <0.1
OD280 nm (1% aq soln) <0.1
pH (1% aq soln) 6 - 7

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty