Luria Bertani Broth, Miller, HiMedia Laboratories
Supplier: PAESEL+LOREI GMBH&CO
M1245-5KG M1245-10KG M1245-2.5KG M1245-25KG
89135-066EA 924.07 USD InStock
89135-066 89134-848 89132-782 71009-262
Luria Bertani Broth, Miller, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Luria Bertani Broth, Miller is used for the cultivation and maintenance of recombinant strains of Escherichia coli and for routine cultivation of fastidious microorganisms.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty