Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cefsulodin sodium salt

Supplier: E. Becker & Co. GmbH

0219867780 0219867783
IC0219867780EA 81.79 USD InStock
IC0219867780 IC0219867783
Cefsulodin sodium salt
Cefsulodin sodium salt
Cefsulodin is a third generation cephalosporin antibiotic that has very specific activity against Pseudomonas aeruginosa. It has no significant activity against other Gram-negative bacteria and very limited activity against Gram-positive bacteria and anaerobic bacteria.
Cefsulodin is a cephalosporin. Cephalosporins disrupt cell wall synthesis by inhibiting PBP crosslinking of peptidoglycan.

Cefuslodin is most commonly used in CIN (cefsulodin-Irgasan-novobiocin) agar to select for Yersinia microorganisms and microorganisms such as E. coli.
Cefsulodin is a substrate for rat Oatp1a4. It has been used to study multidrug resistance-associated protein 4 in efflux transport of prostaglandin E2 across the mouse blood-brain barrier.

Cefsulodin is used to study the effect of expression, binding, and inhibition of PBP1 and other penicillin-binding proteins (PBPs) on bacterial cell wall mucopeptide synthesis.

Store at +4 °C


Formula: C₂₂H₁₉N₄NaO₈S₂
MW: 554.524 g/mol
MDL Number: MFCD07793338
CAS Number: 52152-93-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31f67d7c472bae033bce9179e1f128e14004419d&vid=31f67d7c472bae033bce9179e1f128e14004419d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Physical Appearance White to pale yellow solid
Solubility Soluble in water (50mg/mL-clear, faintly yellow solution).
Storage Store at +4 °C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty