Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Osmium tetraoxide 2% in aqueous solution

Supplier: EPREDIA
Danger

19152 19173 19153 19172 19193 19192
100504-140EA 90.04 USD InStock
100504-140 100504-146 100504-870 100504-144 100504-154 100504-152
Osmium tetraoxide 2% in aqueous solution
Osmium tetraoxide
The osmium tetraoxide is prepared daily in our plant. It is sealed under dry nitrogen.

  • Exact concentrations and volumes in pre-scored amber-ampoules
  • Available in 2% and 4% concentrations
  • Other concentration by special order


Formula: O₄Os
MW: 254.23 g/mol
Boiling Pt: 100 °C (212 °F)
Melting Pt: 40
Density: 1.15624 g/cm³ (9.649 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011150
CAS Number: 20816-12-0
UN: 3287
ADR: 6.1 III
Merck Index: 13,06962
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty