N-DEX™ 4 mil Disposable Nitrile, Powder-Free, Industrial Grade Glove, Showa
Supplier: VWR
17777-200CS 847.2 USD InStock
17777-200 17777-202 17777-204 17777-208
N-DEX™ 4 mil Disposable Nitrile, Powder-Free, Industrial Grade Glove, Showa
Gloves
Perfect 4 mil disposable nitrile glove for lab and industrial use.
  • 100% Nitrile
  • Made in U.S.
  • Powder-free
100% 4 mil Nitrile gloves for lab and industrial use.
Excellent alternative to standard medical exam gloves.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty