SHOWA 8127 A5 Cut Resistant, Uncoated, Wire-Free Glove, Showa
Supplier: Showa
8127-06 8127-07 8127-08 8127-09 8127-10
47742-314CS 457.15 USD InStock
47742-314 47742-316 47742-318 47742-320 47742-322
SHOWA 8127 A5 Cut Resistant, Uncoated, Wire-Free Glove, Showa
Gloves
High quality yarns combined to reach the A5 cut resistant level.
  • Made in U.S.
  • Launderable
  • Amibdextrous
A great glove for high risk cut hazards. Launderable for longer wear.
Certificaciones: ANSI Cut Level A5.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty