ZING Green Safety Eco Safety Sign, DANGER Electrical Equipment Authorized Personnel Only
Supplier: ZING Enterprises
2120 2120A 2120S 1120 1120A 1120S
76023-624EA 30.57 USD InStock
76023-624 76023-626 76023-628 76024-704 76024-706 76024-708
ZING Green Safety Eco Safety Sign, DANGER Electrical Equipment Authorized Personnel Only
Signs
The sign is available in 2 sizes, 7" x 10", and 10" x 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.
Certificaciones: OSHA 29 CFR 1910.145
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty