ZING Green Safety Eco Safety Sign, CAUTION Hearing Protection Must Be Worn In This Area
Supplier: ZING Enterprises
1151S 2151 2151A 2151S 1151 1151A
76022-830EA 19.6 USD InStock
76022-830 76023-774 76023-776 76023-778 76024-878 76024-880
ZING Green Safety Eco Safety Sign, CAUTION Hearing Protection Must Be Worn In This Area
Signs
The sign is available in 2 sizes, 7" x 10", and 10" x 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.
Certificaciones: OSHA 29 CFR 1910.145
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty