ZING Green Safety Industrial Can Crusher
Supplier: ZING Enterprises
7006 7007 7008
76026-006EA 121.71 USD InStock
76026-006 76026-008 76026-010
ZING Green Safety Industrial Can Crusher
Signs
Industrial Can Crusher is the ideal solution to aluminum can management.
  • Made in USA
  • Can management
  • High quality
The can crusher reduces can size which allows more space for more cans resulting in less time spent emptying can collection containers and spending less money on collection bags.
The crusher features a patented pivoting mechanical design that requires 75% less effort than other can crushers. Crushed cans are automatically dropped into an optional collection barrel (sold separately) that holds 48 cans or can be dropped into an existing collection container. The can crusher is made from heavy duty nylon, stainless steel and aluminum and is wall mounted to include mounting hardware. The crusher is designed for 12 oz. aluminum cans and includes a 7" x 10" recycled plastic sign with legend "Please Crush Aluminum Cans". Unit measures 20"H x 6"W x 3-1/2" L. Made in the USA. Optional Can Crusher Barrel Attachment can be ordered using ZING Model Number 7007 or choose the Can Crusher Kit that includes crusher, barrel and sign using ZING Model Number 7008.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty