SHOWA NSK26 Nitrile Supported, Chemical Resistant, 26 Glove, Showa
Supplier: Showa
NSK26-09 NSK26-08 NSK26-10 NSK26-11
89208-338CS 2575.73 USD InStock
89208-338 89208-336 89208-340 89208-342
SHOWA NSK26 Nitrile Supported, Chemical Resistant, 26 Glove, Showa
Gloves
Double dipped Nitrile coating and rough finish provide excellent grip, and protection from chemicals.
  • Rough grip
  • 26" length
  • Highly flexible coating
These gloves are used to handle a wide array of solvents, oils, and acids. Double dipped coating ensures added protection when working with hazardous chemicals.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty