SHOWA S-TEX300 A4 Cut Resistant, Natural Rubber Coated, Kevlar/Hagane Steel Glove, Showa
Supplier: Showa
S-TEX300S-07 S-TEX300M-08 S-TEX300L-09 S-TEX300XL-10
10852-318CS 1543.11 USD InStock
10852-318 10852-320 10852-322 10852-324
SHOWA S-TEX300 A4 Cut Resistant, Natural Rubber Coated, Kevlar/Hagane Steel Glove, Showa
Gloves
Unique Hagane steel component makes glove extremely cut resistant while maintaining flexibility.
  • Hagane steel reinforced Kevlar construction
  • Tough natural rubber coating
These gloves are used in a wide range of cut resistant applications.
Certificaciones: Meets ANSI Level A4 cut level.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty