Brewer Thioglycollate Medium, Modified, Dehydrated Culture Media, HiMedia Laboratories
Supplier: PAESEL+LOREI GMBH&CO
M195A-500G M195-500G
95021-476EA 230.85 USD InStock
95021-476 95021-474
Brewer Thioglycollate Medium, Modified, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For testing the sterility of biological products and for the isolation of aerobic and anaerobic organisms.
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty