Dey-Engley Neutralizing Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: PAESEL+LOREI GMBH&CO
M1062-500G M1062-2.5KG M1062-5KG
95020-396EA 293.28 USD InStock
95020-396 89222-782 89222-784
Dey-Engley Neutralizing Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Dey-Engley Neutralizing Broth is used in disinfectant testing where neutralization of the antiseptics and disinfectants is important for determining its bactericidal activity.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty