KF Streptococcus Agar Base with BCP, Dehydrated Culture Media, HiMedia Laboratories
Supplier: PAESEL+LOREI GMBH&CO
M1007-500G M1007-2.5KG M1007-5KG
95020-274EA 294.61 USD InStock
95020-274 89222-588 89222-590
KF Streptococcus Agar Base with BCP, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the detection and enumeration of fecal streptococci.
  • Microbiological culture media
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty