Kanamycin Esculin Azide Agar Dehydrated Culture Media, HiMedia
Supplier: PAESEL+LOREI GMBH&CO
M510A-500G M510-500G
95021-922EA 360.46 USD InStock
95021-922 95021-920
Kanamycin Esculin Azide Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and identification of Group D streptococci in foodstuffs.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty