Contact Plates , SDA with Lecithin and Polysorbate 80, BD Diagnostics
Supplier: Seca Med.Waagen u.Meßsysteme
292351 292653 215282 292337 215270
71003-782PK 48.67 USD InStock
71003-782 90007-764 90007-922 71003-780 90001-268 90004-231 71003-756
Contact Plates , SDA with Lecithin and Polysorbate 80, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
The BD RODAC™ Snap Lid was designed by environmental monitoring professionals to provide additional security during sampling and transport from a controlled environment to the laboratory.
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty