Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-L-Selectin (CD62L) Rat Monoclonal Antibody [clone: Mel-14]

Supplier: Bio X Cell
BE0021-100MG BE0021-50MG BE0021-5MG BE0021-25MG BE0021-1MG
103252-350EA 3920 USD InStock
103252-350 103252-352 103252-354 103252-356 103572-442 76554-430 76554-730 76554-918 76556-284 76557-360
Anti-L-Selectin (CD62L) Rat Monoclonal Antibody [clone: Mel-14]
Antibodies
InVivoMab anti-mouse L-Selectin (CD62L), Clone: Mel-14, Isotype: rat IgG2a, κ, Applications: In vivo CD62L neutralization

  • Recommended Dilution Buffer: IP0065

Type: Primary
Antigen: L-Selectin (CD62L)
Clonality: Monoclonal
Clone: Mel-14
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty