Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD40L (CD154) Armenian Hamster Monoclonal Antibody [clone: MR-1]

Supplier: Bio X Cell
BE0017-1-1MG BE0017-1-5MG BE0017-1-25MG BE0017-1-100MG BE0017-1-50MG
103252-310EA 3920 USD InStock
103252-310 103252-312 103252-314 103252-316 103572-432 76554-962 76556-156 76557-054 76557-094 76557-172
Anti-CD40L (CD154) Armenian Hamster Monoclonal Antibody [clone: MR-1]
Antibodies
InVivoMab anti-mouse CD154 (CD40L), Clone: MR-1, Isotype: Armenian hamster IgG, Applications: in vivo blocking of CD40/CD40L signaling, in vitro blocking of CD40/CD40L signaling

  • Recommended Dilution Buffer: IP0070

Type: Primary
Antigen: CD40L (CD154)
Clonality: Monoclonal
Clone: MR-1
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty