Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD4 Rat Monoclonal Antibody [clone: YTS 177]

Supplier: Bio X Cell
BE0003-3-25MG BE0003-3-100MG BE0003-3-50MG BE0003-3-5MG
103252-190EA 3920 USD InStock
103252-190 103252-192 103252-194 103252-196 76554-468 76555-590 76556-878 76557-156
Anti-CD4 Rat Monoclonal Antibody [clone: YTS 177]
Antibodies
InVivoMab anti-mouse CD4, Clone: YTS 177, Isotype: rat IgG2a, Applications: in vivo blockade of CD4+ T-cell responses, Western blot

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: YTS 177
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty