Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Armenian Hamster IgG Isotype Control [clone: Polyclonal Armenian]

Supplier: Bio X Cell
BP0091-50MG BP0091-25MG BP0091-100MG BP0091-5MG
103253-328EA 5100 USD InStock
103253-328 103253-330 103253-332 103253-334 76554-426 76554-836 76554-986 76556-622
Armenian Hamster IgG Isotype Control [clone: Polyclonal Armenian]
Antibodies
InVivoPlus polyclonal Armenian hamster IgG, Clone: Polyclonal Armenian, Isotype: Armenian hamster IgG, Applications: in vivo administration

  • Recommended Dilution Buffer: IP0070

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: Polyclonal Armenian
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty