Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Syrian Hamster IgG Isotype Control [clone: Polyclonal Syrian]

Supplier: Bio X Cell
BP0087-50MG BP0087-5MG BP0087-25MG BP0087-100MG
103253-392EA 5100 USD InStock
103253-392 103253-394 103253-396 103253-398 76554-926 76555-616 76556-912 76557-194
Syrian Hamster IgG Isotype Control [clone: Polyclonal Syrian]
Antibodies
InVivoPlus polyclonal Syrian hamster IgG, Clone: Polyclonal Syrian, Isotype: Syrian hamster IgG, Applications: in vivo administration, Functional assays, Immunohistochemistry (frozen), Immunohistochemistry (paraffin), Western blot, Immunofluorescence, Flow Cytometry

  • Recommended Dilution Buffer: IP0070

Recommended Dilution Buffer: IP0070

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: Polyclonal Syrian
Conjugation:
Epitope:
Host: Syrian Hamster
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty