Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-BTLA (CD272) Rat Monoclonal Antibody [clone: PK18.6]

Supplier: Bio X Cell
BE0153-50MG BE0153-1MG BE0153-5MG BE0153-25MG BE0153-100MG
103252-856EA 3920 USD InStock
103252-856 103252-858 103252-860 103252-862 103572-836 76554-348 76554-770 76555-732 76556-558 76556-656
Anti-BTLA (CD272) Rat Monoclonal Antibody [clone: PK18.6]
Antibodies
InVivoMab anti-mouse BTLA (CD272), Clone: PK18.6, Isotype: rat IgG1k, Applications: in vivo stimulation of BTLA, in vitro stimulation of BTLA, Flow cytometry

Type: Primary
Antigen: BTLA (CD272)
Clonality: Monoclonal
Clone: PK18.6
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1k
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty